Assay Details
| Gene Symbol: DTX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126443.5 | NP_178491.1 | 5 - 6 | 1449 | 111 | |
| GenBank mRNA | AK227153.1 | - | 5 - 6 | 1381 | 111 | |
| BT022022.1 | - | 5 - 6 | 1388 | 111 | ||
| BT026371.1 | - | 5 - 6 | 1379 | 111 | ||
More Information
Set Membership: |
|

DTX1
Genomic Map