Assay Details
| Gene Symbol: PCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128510.3 | NP_180517.1 | 1 - 2 | 472 | 152 | |
| GenBank mRNA | BT029466.1 | - | 1 - 2 | 317 | 152 | |
| EU072918.1 | - | 1 - 2 | 317 | 152 | ||
More Information
Set Membership: |
|

PCNA2
Genomic Map