Assay Details
| Gene Symbol: ERF1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036952.1 | NP_001032029.1 | 2 | 1331 | 67 | |
| NM_124162.4 | NP_199599.1 | 2 | 1356 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF1-1| Gene Symbol: ERF1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036952.1 | NP_001032029.1 | 2 | 1331 | 67 | |
| NM_124162.4 | NP_199599.1 | 2 | 1356 | 67 | ||
Set Membership: |
|