Assay Details
| Gene Symbol: DPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342862.1 | NP_001331850.1 | 1 - 2 | 712 | 73 | |
| NM_120707.2 | NP_196243.2 | 1 - 2 | 632 | 73 | ||
More Information
Set Membership: |
|

DPA4
Genomic Map