Assay Details
| Gene Symbol: NHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120715.3 | NP_196250.1 | 1 | 646 | 93 | |
| GenBank mRNA | BT000871.1 | - | 1 | 589 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NHL3| Gene Symbol: NHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120715.3 | NP_196250.1 | 1 | 646 | 93 | |
| GenBank mRNA | BT000871.1 | - | 1 | 589 | 93 | |
Set Membership: |
|