Assay Details
| Gene Symbol: AGL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125811.1 | NP_001119283.1 | 1 | 41 | 92 | |
| NM_122591.3 | NP_198061.2 | 1 | 146 | 92 | ||
| GenBank mRNA | AY141218.1 | - | 1 | 41 | 92 | |
More Information
Set Membership: |
|

AGL54
Genomic Map