Assay Details
| Gene Symbol: NF-YB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124138.2 | NP_199575.1 | 1 | 650 | 75 | |
| GenBank mRNA | AF385744.1 | - | 1 | 650 | 75 | |
| AY078026.1 | - | 1 | 463 | 75 | ||
More Information
Set Membership: |
|

NF-YB2
Genomic Map