Assay Details
Gene Symbol: AT1G61688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036141.4 | NP_001031218.2 | 1 - 2 | 156 | 61 | |
GenBank mRNA | DQ487453.1 | - | 1 - 2 | 121 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
