Assay Details
| Gene Symbol: AT2G39518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084554.2 | NP_001078023.1 | 1 - 2 | 386 | 97 | |
| GenBank mRNA | AK221802.1 | - | 1 - 2 | 235 | 97 | |
More Information
Set Membership: |
|

AT2G39518
Genomic Map