Assay Details
| Gene Symbol: SYP112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335591.1 | NP_001318245.1 | 3 | 1166 | 128 | |
| NM_127384.4 | NP_179418.2 | 2 | 938 | 128 | ||
| GenBank mRNA | DQ446518.1 | - | 1 | 852 | 128 | |
More Information
Set Membership: |
|

SYP112
Genomic Map