Assay Details
| Gene Symbol: AT2G36330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336594.1 | NP_001324366.1 | 1 - 2 | 635 | 65 | |
| NM_129190.3 | NP_181174.2 | 1 - 2 | 635 | 65 | ||
| GenBank mRNA | BT002934.1 | - | 1 - 2 | 613 | 65 | |
More Information
Set Membership: |
|

AT2G36330
Genomic Map