Assay Details
| Gene Symbol: AT2G36330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129190.3 | NP_181174.2 | 2 - 3 | 775 | 71 | |
| GenBank mRNA | BT002934.1 | - | 2 - 3 | 753 | 71 | |
More Information
Set Membership: |
|

AT2G36330
Genomic Map