Assay Details
| Gene Symbol: RTFL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084625.2 | NP_001078094.1 | 1 | 286 | 86 | |
| GenBank mRNA | AY254318.1 | - | 1 | 79 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTFL19| Gene Symbol: RTFL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084625.2 | NP_001078094.1 | 1 | 286 | 86 | |
| GenBank mRNA | AY254318.1 | - | 1 | 79 | 86 | |
Set Membership: |
|