Assay Details
| Gene Symbol: AT4G21745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084952.2 | NP_001078421.1 | 4 - 5 | 601 | 148 | |
| GenBank mRNA | AK230339.1 | - | 4 - 5 | 589 | 148 | |
More Information
Set Membership: |
|

AT4G21745
Genomic Map