Assay Details
| Gene Symbol: NF-YB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085258.1 | NP_001078727.1 | 1 | 746 | 90 | |
| NM_001344740.1 | NP_001318754.1 | 3 | 712 | 90 | ||
| NM_124141.4 | NP_199578.2 | 2 | 841 | 90 | ||
| GenBank mRNA | AY138461.1 | - | 2 | 558 | 90 | |
More Information
Set Membership: |
|

NF-YB6
Genomic Map