Assay Details
| Gene Symbol: AT5G60945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085308.2 | NP_001078777.1 | 1 | 201 | 165 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G60945| Gene Symbol: AT5G60945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085308.2 | NP_001078777.1 | 1 | 201 | 165 | |
Set Membership: |
|