Assay Details
| Gene Symbol: AGL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085329.1 | NP_001078798.1 | 4 - 5 | 532 | 147 | |
| NM_001126026.2 | NP_001119498.1 | 4 - 5 | 567 | 147 | ||
| NM_001345661.1 | NP_001332141.1 | 3 - 4 | 2648 | 147 | ||
| GenBank mRNA | AF312667.1 | - | 4 - 5 | 543 | 147 | |
| AY141211.1 | - | 4 - 5 | 406 | 147 | ||
| AY231441.1 | - | 4 - 5 | 567 | 147 | ||
More Information
Set Membership: |
|

AGL31
Genomic Map