Assay Details
| Gene Symbol: CAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127758.3 | NP_179780.1 | 4 | 1079 | 122 | |
| GenBank mRNA | AY302078.1 | - | 4 | 1048 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAD3| Gene Symbol: CAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127758.3 | NP_179780.1 | 4 | 1079 | 122 | |
| GenBank mRNA | AY302078.1 | - | 4 | 1048 | 122 | |
Set Membership: |
|