Assay Details
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174815.2 | NP_777240.1 | 2 - 3 | 364 | 148 | |
| GenBank mRNA | BC105363.1 | - | 2 - 3 | 364 | 148 | |
| M95793.1 | - | 2 - 3 | 336 | 148 | ||
More Information
Set Membership: |
|

YBX1
Genomic Map