Assay Details
| Gene Symbol: PANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075463.1 | NP_001068931.1 | 2 - 3 | 695 | 80 | |
| GenBank mRNA | BC123859.1 | - | 2 - 3 | 695 | 80 | |
More Information
Set Membership: |
|

PANK3
Genomic Map