Assay Details
| Gene Symbol: SPOCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101115.1 | NP_001094585.1 | 3 - 4 | 455 | 81 | |
| GenBank mRNA | BC151268.1 | - | 3 - 4 | 455 | 81 | |
More Information
Set Membership: |
|

SPOCK2
Genomic Map