Assay Details
| Gene Symbol: NKX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191458.1 | NP_001178387.1 | 1 - 2 | 796 | 95 | |
| GenBank mRNA | EU293622.1 | - | 1 - 2 | 122 | 95 | |
More Information
Set Membership: |
|

NKX2-1
Genomic Map