Assay Details
| Gene Symbol: TBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076290.2 | NP_001069758.1 | 3 - 4 | 389 | 77 | |
| GenBank mRNA | BC109666.1 | - | 3 - 4 | 390 | 77 | |
| EU140625.1 | - | 3 - 4 | 449 | 77 | ||
More Information
Set Membership: |
|

TBPL1
Genomic Map