Assay Details
| Gene Symbol: ZNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035268.2 | NP_001030345.1 | 6 - 7 | 751 | 61 | |
| GenBank mRNA | BC107549.1 | - | 6 - 7 | 752 | 61 | |
More Information
Set Membership: |
|

ZNF207
Genomic Map