Assay Details
| Gene Symbol: SLC22A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130755.2 | NP_001124227.1 | 7 - 8 | 1435 | 82 | |
| GenBank mRNA | BC102752.2 | - | 6 - 7 | 1108 | 82 | |
More Information
Set Membership: |
|

SLC22A17
Genomic Map