Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113281.1 | NP_001106752.1 | 2 - 3 | 1537 | 67 | |
GenBank mRNA | DN524278.1 | - | 2 - 3 | 646 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
