Assay Details
| Gene Symbol: CDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014934.1 | NP_001014934.1 | 6 - 7 | 995 | 66 | |
| GenBank mRNA | BC150026.1 | - | 6 - 7 | 965 | 66 | |
| BT020790.1 | - | 6 - 7 | 995 | 66 | ||
More Information
Set Membership: |
|

CDK2
Genomic Map