Assay Details
| Gene Symbol: MEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076161.3 | NP_001069629.1 | 3 - 4 | 728 | 70 | |
| GenBank mRNA | BC120005.1 | - | 3 - 4 | 695 | 70 | |
More Information
Set Membership: |
|

MEN1
Genomic Map