Assay Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033121.1 | NP_001028293.1 | 3 - 4 | 551 | 139 | |
| GenBank mRNA | DQ162621.1 | - | 3 - 4 | 551 | 139 | |
More Information
Set Membership: |
|

ZNF148
Genomic Map