Assay Details
| Gene Symbol: PCGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077980.1 | NP_001071448.1 | 8 - 9 | 900 | 66 | |
| GenBank mRNA | BC126565.1 | - | 8 - 9 | 900 | 66 | |
More Information
Set Membership: |
|

PCGF5
Genomic Map