Assay Details
| Gene Symbol: NONO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046554.1 | NP_001040019.1 | 4 - 5 | 697 | 64 | |
| GenBank mRNA | BC105532.1 | - | 4 - 5 | 697 | 64 | |
More Information
Set Membership: |
|

NONO
Genomic Map