Assay Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077953.1 | NP_001071421.1 | 2 - 3 | 503 | 74 | |
| GenBank mRNA | BC123728.1 | - | 2 - 3 | 503 | 74 | |
More Information
Set Membership: |
|

RNF11
Genomic Map