Assay Details
| Gene Symbol: CELF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099068.2 | NP_001092538.1 | 11 - 12 | 1396 | 57 | |
| GenBank mRNA | BC146142.1 | - | 11 - 12 | 1397 | 57 | |
More Information
Set Membership: |
|

CELF4
Genomic Map