Assay Details
| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099108.2 | NP_001092578.1 | 8 - 9 | 942 | 54 | |
| GenBank mRNA | BC146137.1 | - | 8 - 9 | 943 | 54 | |
More Information
Set Membership: |
|

PPP4C
Genomic Map