Assay Details
| Gene Symbol: MED17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034730.2 | NP_001029902.2 | 2 - 3 | 531 | 155 | |
| GenBank mRNA | BC103341.1 | - | 2 - 3 | 565 | 155 | |
| BT021158.1 | - | 2 - 3 | 533 | 155 | ||
More Information
Set Membership: |
|

MED17
Genomic Map