Assay Details
| Gene Symbol: PCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034711.1 | NP_001029883.1 | 9 - 10 | 745 | 60 | |
| GenBank mRNA | BC103397.1 | - | 9 - 10 | 745 | 60 | |
More Information
Set Membership: |
|

PCBP2
Genomic Map