Assay Details
| Gene Symbol: PDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100331.1 | NP_001093801.1 | 6 - 7 | 741 | 98 | |
| GenBank mRNA | BC149261.1 | - | 6 - 7 | 741 | 98 | |
| KC787698.1 | - | 6 - 7 | 740 | 98 | ||
More Information
Set Membership: |
|

PDSS1
Genomic Map