Assay Details
| Gene Symbol: IRAK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075998.1 | NP_001069466.1 | 6 - 7 | 794 | 136 | |
| GenBank mRNA | BC114720.1 | - | 6 - 7 | 794 | 136 | |
| EU528625.1 | - | 6 - 7 | 970 | 136 | ||
| EU528626.1 | - | 6 - 7 | 812 | 136 | ||
More Information
Set Membership: |
|

IRAK4
Genomic Map