Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035451.1 | NP_001030528.1 | 5 - 6 | 494 | 59 | |
| GenBank mRNA | BC104608.2 | - | 5 - 6 | 494 | 59 | |
More Information
Set Membership: |
|

TMEM222
Genomic Map