Assay Details
| Gene Symbol: FYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077972.1 | NP_001071440.1 | 8 - 9 | 1183 | 91 | |
| GenBank mRNA | BC126591.1 | - | 8 - 9 | 1183 | 91 | |
More Information
Set Membership: |
|

FYN
Genomic Map