Assay Details
| Gene Symbol: PPP2R5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083468.1 | NP_001076937.1 | 2 - 3 | 681 | 129 | |
| GenBank mRNA | BC123807.1 | - | 2 - 3 | 681 | 129 | |
More Information
Set Membership: |
|

PPP2R5E
Genomic Map