Assay Details
| Gene Symbol: SRSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078126.2 | NP_001071594.1 | 5 - 6 | 571 | 66 | |
| GenBank mRNA | BC103100.1 | - | 5 - 6 | 573 | 66 | |
More Information
Set Membership: |
|

SRSF10
Genomic Map