Assay Details
| Gene Symbol: PARVA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099144.1 | NP_001092614.1 | 6 - 7 | 715 | 63 | |
| GenBank mRNA | BC142362.1 | - | 6 - 7 | 715 | 63 | |
More Information
Set Membership: |
|

PARVA
Genomic Map