Assay Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191183.1 | NP_001178112.1 | 2 - 3 | 482 | 62 | |
| GenBank mRNA | BC114909.1 | - | 2 - 3 | 463 | 62 | |
More Information
Set Membership: |
|

ACBD3
Genomic Map