Assay Details
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034719.2 | NP_001029891.1 | 1 - 2 | 78 | 93 | |
| GenBank mRNA | BC102210.1 | - | 1 - 2 | 80 | 93 | |
More Information
Set Membership: |
|

OTUB1
Genomic Map