Assay Details
| Gene Symbol: RAB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099094.2 | NP_001092564.1 | 2 - 3 | 443 | 71 | |
| GenBank mRNA | BC142252.1 | - | 2 - 3 | 444 | 71 | |
More Information
Set Membership: |
|

RAB29
Genomic Map