Assay Details
| Gene Symbol: AMFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045974.1 | NP_001039439.1 | 8 - 9 | 1181 | 62 | |
| GenBank mRNA | BC112457.1 | - | 8 - 9 | 1181 | 62 | |
More Information
Set Membership: |
|

AMFR
Genomic Map