Assay Details
| Gene Symbol: RNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075720.2 | NP_001069188.1 | 1 - 2 | 176 | 74 | |
| GenBank mRNA | BC120122.1 | - | 1 - 2 | 177 | 74 | |
More Information
Set Membership: |
|

RNF7
Genomic Map