Assay Details
| Gene Symbol: PRR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035396.1 | NP_001030473.1 | 10 - 11 | 1527 | 123 | |
| GenBank mRNA | BC120423.1 | - | 10 - 11 | 1530 | 123 | |
| BT021795.1 | - | 10 - 11 | 1527 | 123 | ||
More Information
Set Membership: |
|

PRR14
Genomic Map