Assay Details
| Gene Symbol: FBXO32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046155.1 | NP_001039620.1 | 2 - 3 | 556 | 101 | |
| GenBank mRNA | BC112888.1 | - | 2 - 3 | 556 | 101 | |
More Information
Set Membership: |
|

FBXO32
Genomic Map